5-Brom-3-chlor-2-fluorphenol

Lieferant: COMBI-BLOCKS

HC-3520-1G
COBBHC-3520-1GEA 236 EUR
COBBHC-3520-1G
5-Brom-3-chlor-2-fluorphenol
5-Brom-3-chlor-2-fluorphenol
Formel: C₆H₃BrClFO
Molecular Weight: 225,4428 g/mol
MDL: MFCD18914516
CAS-Nummer: 1305322-97-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR