3,5-Dimethyl-4-iodoaniline

Lieferant: COMBI-BLOCKS

Synonyme: 4-Iodo-3,5-dimethylaniline

HC-4054-1G
COBBHC-4054-1GEA 65.8 EUR
COBBHC-4054-1G
3,5-Dimethyl-4-iodoaniline
3,5-Dimethyl-4-iodoaniline
Formel: C₈H₁₀IN
Molecular Weight: 247,0800 g/mol
MDL: MFCD06664471
CAS-Nummer: 117832-15-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR