Isochinolin-8-amin
Lieferant: COMBI-BLOCKS
Synonyme:
Isoquinolin-8-amine, 8-Aminoisoquinoline
HC-6001-1G
HC-6001-5G
COBBHC-6001-1GEA
0
EUR
COBBHC-6001-1G
COBBHC-6001-5G
Isochinolin-8-amin
Isochinolin-8-amin
Formel:
C₉H₈N₂ |
MDL:
MFCD00179553 CAS-Nummer: 23687-27-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...