6,7-Dimethoxyisochinolin
Lieferant: COMBI-BLOCKS
HC-6011-5G
HC-6011-1G
COBBHC-6011-5GEA
1090
EUR
COBBHC-6011-5G
COBBHC-6011-1G
6,7-Dimethoxyisochinolin
6,7-Dimethoxyisoquinoline
Formel:
C₁₁H₁₁NO₂ |
MDL:
MFCD00666134 CAS-Nummer: 15248-39-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...