6,7-Dimethoxyisochinolin

Lieferant: COMBI-BLOCKS

HC-6011-5G HC-6011-1G
COBBHC-6011-5GEA 1090 EUR
COBBHC-6011-5G COBBHC-6011-1G
6,7-Dimethoxyisochinolin
6,7-Dimethoxyisoquinoline
Formel: C₁₁H₁₁NO₂
MDL: MFCD00666134
CAS-Nummer: 15248-39-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR