8-Isochinolinol

Lieferant: COMBI-BLOCKS

Synonyme: 8-Hydroxyisoquinoline, Isoquinolin-8-ol

HC-6024-1G
COBBHC-6024-1GEA 208 EUR
COBBHC-6024-1G
8-Isochinolinol
8-Isochinolinol
Formel: C₉H₇NO
Molecular Weight: 145,1600 g/mol
MDL: MFCD00661583
CAS-Nummer: 3482-14-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR