7-Methoxyisochinolin
Lieferant: COMBI-BLOCKS
HC-6026-5G
HC-6026-1G
COBBHC-6026-5GEA
0
EUR
COBBHC-6026-5G
COBBHC-6026-1G
7-Methoxyisochinolin
7-Methoxyisoquinoline
Formel:
C₁₀H₉NO |
MDL:
MFCD09030747 CAS-Nummer: 39989-39-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...