1,3-Dichlor-7-fluorisochinolin

Lieferant: COMBI-BLOCKS

HC-6033-1G
COBBHC-6033-1GEA 377 EUR
COBBHC-6033-1G
1,3-Dichlor-7-fluorisochinolin
1,3-Dichloro-7-fluoroisoquinoline
Formel: C₉H₄Cl₂FN
Molecular Weight: 216.0392 g/mol
MDL: MFCD09763685
CAS-Nummer: 941294-25-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR