8-Methyl-4-chinazolon

Lieferant: COMBI-BLOCKS

Synonyme: 8-Methylquinazolin-4(3H)-one

HC-6039-1G
COBBHC-6039-1GEA 47.4 EUR
COBBHC-6039-1G
8-Methyl-4-chinazolon
8-Methyl-4-chinazolon
Formel: C₉H₈N₂O
Molecular Weight: 160,1726 g/mol
MDL: MFCD01685945
CAS-Nummer: 19181-54-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR