6-Chlorimidazo[1,2-a]pyridin Hydrochlorid

Lieferant: COMBI-BLOCKS

HC-6080-25G HC-6080-1G HC-6080-5G
COBBHC-6080-25GEA 0 EUR
COBBHC-6080-25G COBBHC-6080-1G COBBHC-6080-5G
6-Chlorimidazo[1,2-a]pyridin Hydrochlorid
6-Chlorimidazo[1,2-a]pyridin Hydrochlorid
Formel: C₇H₆Cl₂N₂
MDL: MFCD09475876
CAS-Nummer: 957035-24-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR