6-Nitroimidazo[1,2-a]pyridin Hydrochlorid

Lieferant: COMBI-BLOCKS

HC-6160-1G HC-6160-25G HC-6160-5G
COBBHC-6160-1GEA 0 EUR
COBBHC-6160-1G COBBHC-6160-25G COBBHC-6160-5G
6-Nitroimidazo[1,2-a]pyridin Hydrochlorid
6-Nitroimidazo[1,2-a]pyridin Hydrochlorid
Formel: C₇H₆ClN₃O₂
MDL: MFCD09800971
CAS-Nummer: 957120-37-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR