Ethyl-8-brom-4-hydroxychinolin-3-carboxylat

Lieferant: COMBI-BLOCKS

Synonyme: Ethyl 8-bromo-4-hydroxyquinoline-3-carboxylate

HC-6213-1G
COBBHC-6213-1GEA 112 EUR
COBBHC-6213-1G
Ethyl-8-brom-4-hydroxychinolin-3-carboxylat
Ethyl-8-brom-4-hydroxychinolin-3-carboxylat
Formel: C₁₂H₁₀BrNO₃
Molecular Weight: 296,1167 g/mol
MDL: MFCD00173380
CAS-Nummer: 35975-57-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR