Ethyl-6,8-difluor-4-hydroxychinolin-3-carboxylat

Lieferant: COMBI-BLOCKS

HC-6215-1G
COBBHC-6215-1GEA 303 EUR
COBBHC-6215-1G
Ethyl-6,8-difluor-4-hydroxychinolin-3-carboxylat
Ethyl-6,8-difluor-4-hydroxychinolin-3-carboxylat
Formel: C₁₂H₉F₂NO
MDL: MFCD00173349
CAS-Nummer: 107555-38-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR