4-(3-Brom-5-(trifluormethyl)phenylsulfonyl)morpholin

Lieferant: COMBI-BLOCKS

HC-6301-5G HC-6301-1G
COBBHC-6301-5GEA 0 EUR
COBBHC-6301-5G COBBHC-6301-1G
4-(3-Brom-5-(trifluormethyl)phenylsulfonyl)morpholin
4-(3-Brom-5-(trifluormethyl)phenylsulfonyl)morpholin
Formel: C₁₁H₁₁BrF₃NO₃S
MDL: MFCD09801007
CAS-Nummer: 951884-77-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR