3-Brom-N-ethyl-5-methylbenzolsulfonamid

Lieferant: COMBI-BLOCKS

HC-6325-1G
COBBHC-6325-1GEA 149 EUR
COBBHC-6325-1G
3-Brom-N-ethyl-5-methylbenzolsulfonamid
3-Brom-N-ethyl-5-methylbenzolsulfonamid
Formel: C₉H₁₂BrNO₂S
MDL: MFCD09801020
CAS-Nummer: 951885-52-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR