Ethyl 4-oxo-7-(trifluoromethyl)-1,4-dihydroquinoline-3-carboxylate

Lieferant: COMBI-BLOCKS

HC-6334-1G
COBBHC-6334-1GEA 193 EUR
COBBHC-6334-1G
Ethyl 4-oxo-7-(trifluoromethyl)-1,4-dihydroquinoline-3-carboxylate
Ethyl 4-oxo-7-(trifluoromethyl)-1,4-dihydroquinoline-3-carboxylate
Formel: C₁₃H₁₀F₃NO₃
Molecular Weight: 285.22 g/mol
MDL: MFCD01080777
CAS-Nummer: 71083-04-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR