Ethyl-4-hydroxy-8-(trifluormethyl)chinolin-3-carboxylat
Lieferant: COMBI-BLOCKS
Synonyme:
Ethyl 4-hydroxy-8-(trifluoromethyl)quinoline-3-carboxylate
HC-6355-5G
HC-6355-1G
COBBHC-6355-5GEA
0
EUR
COBBHC-6355-5G
COBBHC-6355-1G
Ethyl-4-hydroxy-8-(trifluormethyl)chinolin-3-carboxylat
Ethyl-4-hydroxy-8-(trifluormethyl)chinolin-3-carboxylat
Formel:
C₁₃H₁₀F₃NO₃ |
MDL:
MFCD09878378 CAS-Nummer: 23851-84-5 EINECS: 245-914-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...