2-Brom-N-cyclopropyl-4-(trifluormethoxy)benzolsulfonamid

Lieferant: COMBI-BLOCKS

HC-6361-1G
COBBHC-6361-1GEA 204 EUR
COBBHC-6361-1G
2-Brom-N-cyclopropyl-4-(trifluormethoxy)benzolsulfonamid
2-Brom-N-cyclopropyl-4-(trifluormethoxy)benzolsulfonamid
Formel: C₁₀H₉BrF₃NO₃S
MDL: MFCD09878380
CAS-Nummer: 957062-79-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR