2-Brom-6-(4-morpholino)pyridin

Lieferant: COMBI-BLOCKS

HC-6366-5G HC-6366-25G HC-6366-1G
COBBHC-6366-5GEA 0 EUR
COBBHC-6366-5G COBBHC-6366-25G COBBHC-6366-1G
2-Brom-6-(4-morpholino)pyridin
2-Brom-6-(4-morpholino)pyridin
Formel: C₉H₁₁BrN₂O
MDL: MFCD07772824
CAS-Nummer: 332134-60-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR