3-Brom-N-tert-butyl-5-methylbenzolsulfonamid

Lieferant: COMBI-BLOCKS

HC-6457-5G HC-6457-1G
COBBHC-6457-5GEA 451 EUR
COBBHC-6457-5G COBBHC-6457-1G
3-Brom-N-tert-butyl-5-methylbenzolsulfonamid
3-Brom-N-tert-butyl-5-methylbenzolsulfonamid
Formel: C₁₁H₁₆BrNO₂S
MDL: MFCD10699659
CAS-Nummer: 1033201-97-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR