3-Brom-N-tert-butyl-5-methylbenzolsulfonamid
Lieferant: COMBI-BLOCKS
HC-6457-5G
HC-6457-1G
COBBHC-6457-5GEA
451
EUR
COBBHC-6457-5G
COBBHC-6457-1G
3-Brom-N-tert-butyl-5-methylbenzolsulfonamid
3-Brom-N-tert-butyl-5-methylbenzolsulfonamid
Formel:
C₁₁H₁₆BrNO₂S |
MDL:
MFCD10699659 CAS-Nummer: 1033201-97-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...