4-(Pyrrolidin-1-yl)-8-(trifluormethyl)chinolin

Lieferant: COMBI-BLOCKS

HC-6482-1G
COBBHC-6482-1GEA 149 EUR
COBBHC-6482-1G
4-(Pyrrolidin-1-yl)-8-(trifluormethyl)chinolin
4-(Pyrrolidin-1-yl)-8-(trifluormethyl)chinolin
Formel: C₁₄H₁₃F₃N₂
MDL: MFCD09972163
CAS-Nummer: 1020253-07-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR