4-Phenoxypiperidin

Lieferant: COMBI-BLOCKS

HC-6488-5G HC-6488-1G
COBBHC-6488-5GEA 384 EUR
COBBHC-6488-5G COBBHC-6488-1G
4-Phenoxypiperidin
4-Phenoxypiperidin
Formel: C₁₁H₁₅NO
Molecular Weight: 177.25 g/mol
MDL: MFCD04114971
CAS-Nummer: 3202-33-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR