Imidazo[1,2-a]pyridin-3-carbaldehyd

Lieferant: COMBI-BLOCKS

HC-6659-5G HC-6659-1G
COBBHC-6659-5GEA 0 EUR
COBBHC-6659-5G COBBHC-6659-1G
Imidazo[1,2-a]pyridin-3-carbaldehyd
Imidazo[1,2-a]pyridin-3-carbaldehyd
Formel: C₈H₆N₂O
MDL: MFCD07778354
CAS-Nummer: 6188-43-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR