6-Bromimidazo[1,2-a]pyridin-3-carbaldehyd

Lieferant: COMBI-BLOCKS

HC-6660-1G
COBBHC-6660-1GEA 352 EUR
COBBHC-6660-1G
6-Bromimidazo[1,2-a]pyridin-3-carbaldehyd
6-Bromimidazo[1,2-a]pyridin-3-carbaldehyd
Formel: C₈H₅BrN₂O
Molecular Weight: 225,0400 g/mol
MDL: MFCD09864909
CAS-Nummer: 30384-96-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR