6-Chlorimidazo[1,2-a]pyridin-3-carbaldehyd

Lieferant: COMBI-BLOCKS

HC-6661-1G
COBBHC-6661-1GEA 0 EUR
COBBHC-6661-1G
6-Chlorimidazo[1,2-a]pyridin-3-carbaldehyd
6-Chlorimidazo[1,2-a]pyridin-3-carbaldehyd
Formel: C₈H₅ClN₂O
MDL: MFCD09909649
CAS-Nummer: 29096-59-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR