Broxyquinolin
Lieferant: COMBI-BLOCKS
|
Achtung
|
HC-6706-100G
HC-6706-25G
COBBHC-6706-100GEA
0
EUR
COBBHC-6706-100G
COBBHC-6706-25G
Broxyquinolin
Broxyquinolin
Formel:
C₉H₅Br₂NO Siedepunkt: 371 °C (1013 hPa) Dichte: 2,189 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006785 CAS-Nummer: 521-74-4 EINECS: 208-317-8 Merck Index: 13,01438 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...