Ethyl-2-amino-4-methylthiazol-5-carboxylat
Lieferant: COMBI-BLOCKS
Synonyme:
Ethyl 2-amino-4-methylthiazole-5-carboxylate
HC-6710-5G
HC-6710-25G
HC-6710-100G
COBBHC-6710-5GEA
0
EUR
COBBHC-6710-5G
COBBHC-6710-25G
COBBHC-6710-100G
Ethyl-2-amino-4-methylthiazol-5-carboxylat
Ethyl-2-amino-4-methylthiazol-5-carboxylat
Formel:
C₇H₁₀N₂O₂S |
MDL:
MFCD00123414 CAS-Nummer: 7210-76-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...