3-Amino-1H-pyrazol

Lieferant: COMBI-BLOCKS

Synonyme: 3-Pyrazolamine, 3-AP, 1H-Pyrazol-3-amine, 3-Aminopyrazole

HC-6711-100G
COBBHC-6711-100GEA 218 EUR
COBBHC-6711-100G
3-Amino-1H-pyrazol
3-Amino-1H-pyrazole
Formel: C₃H₅N₃
Molecular Weight: 83,09 g/mol
MDL: MFCD00005236
CAS-Nummer: 1820-80-0
EINECS: 217-338-1
UN: 3259
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR