8-Brom-6-methylimidazo[1,2-a]pyridin-3-carbaldehyd

Lieferant: COMBI-BLOCKS

HC-6739-1G
COBBHC-6739-1GEA 0 EUR
COBBHC-6739-1G
8-Brom-6-methylimidazo[1,2-a]pyridin-3-carbaldehyd
8-Brom-6-methylimidazo[1,2-a]pyridin-3-carbaldehyd
Formel: C₉H₇BrN₂O
MDL: MFCD10699669
CAS-Nummer: 1033202-08-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR