6-Brom-7-methylimidazo[1,2-a]pyridin-3-carbaldehyd
Lieferant: COMBI-BLOCKS
HC-6740-1G
COBBHC-6740-1GEA
451
EUR
COBBHC-6740-1G
6-Brom-7-methylimidazo[1,2-a]pyridin-3-carbaldehyd
6-Brom-7-methylimidazo[1,2-a]pyridin-3-carbaldehyd
Formel:
C₉H₇BrN₂O |
MDL:
MFCD11504923 CAS-Nummer: 1072944-75-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...