5-Chlor-8-methyl-2-(trifluormethyl)-4-chinolinol

Lieferant: COMBI-BLOCKS

HC-6753-1G
COBBHC-6753-1GEA 149 EUR
COBBHC-6753-1G
5-Chlor-8-methyl-2-(trifluormethyl)-4-chinolinol
5-Chlor-8-methyl-2-(trifluormethyl)-4-chinolinol
Formel: C₁₁H₇ClF₃NO
MDL: MFCD11053825
CAS-Nummer: 59108-09-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR