2-Acetamido-5-bromthiazol

Lieferant: COMBI-BLOCKS

HC-6857-25G HC-6857-1G HC-6857-5G
COBBHC-6857-25GEA 0 EUR
COBBHC-6857-25G COBBHC-6857-1G COBBHC-6857-5G
2-Acetamido-5-bromthiazol
2-Acetamido-5-bromthiazol
Formel: C₅H₅BrN₂OS
MDL: MFCD00017336
CAS-Nummer: 7336-54-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR