2-Acetamido-5-bromthiazol
Lieferant: COMBI-BLOCKS
Synonyme:
N-(5-bromothiazol-2-yl)acetamide
HC-6857-25G
HC-6857-1G
HC-6857-5G
COBBHC-6857-25GEA
0
EUR
COBBHC-6857-25G
COBBHC-6857-1G
COBBHC-6857-5G
2-Acetamido-5-bromthiazol
2-Acetamido-5-bromthiazol
Formel:
C₅H₅BrN₂OS |
MDL:
MFCD00017336 CAS-Nummer: 7336-54-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...