Methyl-5-brom-4-hydroxy-8-methylchinolin-2-carboxylat

Lieferant: COMBI-BLOCKS

HC-6870-1G
COBBHC-6870-1GEA 0 EUR
COBBHC-6870-1G
Methyl-5-brom-4-hydroxy-8-methylchinolin-2-carboxylat
Methyl-5-brom-4-hydroxy-8-methylchinolin-2-carboxylat
Formel: C₁₂H₁₀BrNO₃
MDL: MFCD11855939
CAS-Nummer: 1133116-21-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR