3-Amino-2-brom-6-chlorpyridin

Lieferant: COMBI-BLOCKS

Synonyme: 2-Brom-6-chlor-3-pyridylamin

HC-6875-5G HC-6875-25G HC-6875-1G
COBBHC-6875-5GEA 323 EUR
COBBHC-6875-5G COBBHC-6875-25G COBBHC-6875-1G
3-Amino-2-brom-6-chlorpyridin
3-Amino-2-bromo-6-chloropyridine
Formel: C₅H₄BrClN₂
Molecular Weight: 207.46 g/mol
MDL: MFCD11504927
CAS-Nummer: 1050501-88-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR