Methyl-6-brom-7-chlor-8-methyl-4-hydroxychinolin-2-carboxylat

Lieferant: COMBI-BLOCKS

HC-6945-1G
COBBHC-6945-1GEA 149 EUR
COBBHC-6945-1G
Methyl-6-brom-7-chlor-8-methyl-4-hydroxychinolin-2-carboxylat
Methyl-6-brom-7-chlor-8-methyl-4-hydroxychinolin-2-carboxylat
Formel: C₁₂H₉BrClNO₃
MDL: MFCD11053780
CAS-Nummer: 1065074-46-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR