2,4,5-Triiodimidazol

Lieferant: COMBI-BLOCKS

Synonyme: 2,4,5-Triiod-1H-imidazol

HI-1003-5G HI-1003-100G HI-1003-25G
COBBHI-1003-5GEA 79.1 EUR
COBBHI-1003-5G COBBHI-1003-100G COBBHI-1003-25G
2,4,5-Triiodimidazol
2,4,5-Triiodoimidazole
Formel: C₃HI₃N₂
Molecular Weight: 445.77 g/mol
MDL: MFCD00068812
CAS-Nummer: 1746-25-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR