4-Iod-1-methylimidazol

Lieferant: COMBI-BLOCKS

HI-1004-1G
COBBHI-1004-1GEA 0 EUR
COBBHI-1004-1G
4-Iod-1-methylimidazol
4-Iodo-1-methylimidazole
Formel: C₄H₅IN₂
MDL: MFCD01632214
CAS-Nummer: 71759-87-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR