2,5-Dibrom-1-methylimidazol
Lieferant: COMBI-BLOCKS
Synonyme:
2,5-Dibrom-1-methyl-1H-imidazol
HI-1010-1G
COBBHI-1010-1GEA
186
EUR
COBBHI-1010-1G
2,5-Dibrom-1-methylimidazol
2,5-Dibromo-1-methylimidazole
Formel:
C₄H₄Br₂N₂ Molecular Weight: 239,9 g/mol |
MDL:
MFCD01632217 CAS-Nummer: 53857-59-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...