4-Iod-1-methylimidazol-5-carbaldehyd

Lieferant: COMBI-BLOCKS

Synonyme: 4-Iodo-1-methyl-1H-imidazole-5-carbaldehyde

HI-1028-1G
COBBHI-1028-1GEA 1580 EUR
COBBHI-1028-1G
4-Iod-1-methylimidazol-5-carbaldehyd
4-Iod-1-methylimidazol-5-carbaldehyd
Formel: C₅H₅IN₂O
Molecular Weight: 236,0105 g/mol
MDL: MFCD01632219
CAS-Nummer: 189014-13-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR