2-Brom-1-tritylimidazol

Lieferant: COMBI-BLOCKS

HI-1044-1G
COBBHI-1044-1GEA 182 EUR
COBBHI-1044-1G
2-Brom-1-tritylimidazol
2-Brom-1-tritylimidazol
Formel: C₂₂H₁₇BrN₂
MDL: MFCD02179528
CAS-Nummer: 67478-47-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR