2-Iod-1-tritylimidazol
Lieferant: COMBI-BLOCKS
HI-1045-1G
COBBHI-1045-1GEA
249
EUR
COBBHI-1045-1G
2-Iod-1-tritylimidazol
2-Iod-1-tritylimidazol
Formel:
C₂₂H₁₇IN₂ Molecular Weight: 436,29 g/mol |
MDL:
MFCD02179529 CAS-Nummer: 67478-46-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...