2-Chlorimidazol

Lieferant: COMBI-BLOCKS

Synonyme: 2-Chlor-1H-imidazol

HI-1046-1G
COBBHI-1046-1GEA 385 EUR
COBBHI-1046-1G
2-Chlorimidazol
2-Chloroimidazole
Formel: C₃H₃ClN₂
Molecular Weight: 102,52 g/mol
MDL: MFCD02179530
CAS-Nummer: 16265-04-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR