2-Chlor-1-methylimidazol
Lieferant: COMBI-BLOCKS
Synonyme:
2-Chlor-1-methyl-1H-imidazol
HI-1049-1G
COBBHI-1049-1GEA
180
EUR
COBBHI-1049-1G
2-Chlor-1-methylimidazol
2-Chloro-1-methylimidazole
Formel:
C₄H₅ClN₂ Molecular Weight: 116,55 g/mol |
MDL:
MFCD02179531 CAS-Nummer: 253453-91-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...