2,5-Diiod-1-methylimidazol
Lieferant: COMBI-BLOCKS
Synonyme:
2,5-Diiod-1-methyl-1H-imidazol
HI-1061-1G
COBBHI-1061-1GEA
431
EUR
COBBHI-1061-1G
2,5-Diiod-1-methylimidazol
2,5-Diiodo-1-methylimidazole
Formel:
C₄H₄I₂N₂ Molecular Weight: 333,9 g/mol |
MDL:
MFCD02179540 CAS-Nummer: 86026-81-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...