(1H-Imidazol-4-yl)methanol Hydrochlorid
Lieferant: COMBI-BLOCKS
Synonyme:
4-Imidazolemethanol hydrochloride, 4(5)-Imidazolemethanol hydrochloride, 4-(Hydroxymethyl)imidazole hydrochloride
HI-1077-25G
COBBHI-1077-25GEA
410
EUR
COBBHI-1077-25G
(1H-Imidazol-4-yl)methanol Hydrochlorid
(1H-Imidazol-4-yl)methanol Hydrochlorid
Formel:
C₄H₇ClN₂O Molecular Weight: 134,57 g/mol |
MDL:
MFCD00012697 CAS-Nummer: 32673-41-9 EINECS: 251-150-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...