4,5-Dicyan-1-methylimidazol

Lieferant: COMBI-BLOCKS

Synonyme: 4,5-Dicyan-1-methyl-1H-imidazol, 1-Methylimidazol-4,5-dicarbonitril

HI-1088-5G
COBBHI-1088-5GEA 49.7 EUR
COBBHI-1088-5G
4,5-Dicyan-1-methylimidazol
4,5-Dicyano-1-methylimidazole
Formel: C₆H₄N₄
Molecular Weight: 132,12 g/mol
MDL: MFCD01567299
CAS-Nummer: 19485-35-9
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR