1-Methylimidazol-4-carbonsäure

Lieferant: COMBI-BLOCKS

Synonyme: 1-Methyl-1H-imidazole-4-carboxylic acid

HI-1090-5G HI-1090-1G HI-1090-25G
COBBHI-1090-5GEA 0 EUR
COBBHI-1090-5G COBBHI-1090-1G COBBHI-1090-25G
1-Methylimidazol-4-carbonsäure
1-Methylimidazol-4-carbonsäure
Formel: C₅H₆N₂O₂
MDL: MFCD02179560
CAS-Nummer: 41716-18-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR