1-Methylimidazol-4-carbonsäuremethylester

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 1-methyl-1H-imidazole-4-carboxylate

HI-1091-1G
COBBHI-1091-1GEA 102 EUR
COBBHI-1091-1G
1-Methylimidazol-4-carbonsäuremethylester
1-Methylimidazol-4-carbonsäuremethylester
Formel: C₆H₈N₂O₂
Molecular Weight: 140,1400 g/mol
MDL: MFCD02179561
CAS-Nummer: 17289-19-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR